
1,4-Bis(4-bromophenyl)-1,3-butadiyne ≥98.0% (by HPLC)
Catalog # TCIAB4013-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
About this item
:
:360.05 g/mol
:GHS07
CAS Number:959-88-6
Formula:C₁₆H₈Br₂
MDL Number:MFCD00825678