N-(tert-Butoxycarbonyl)-L-propargylglycine ≥97.0% (by HPLC, titration analysis)
TCIAB4007-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Melting point:88.0 to 92.0 deg-C
- Specific rotation [a]20/D:+24.0 to +27.0 deg (C=1、MeOH)
:
:213.23 g/mol
:Freezer
:63039-48-5
:C₁₀H₁₅NO₄
:MFCD01320855