4-[N,N-Bis(4-bromophenyl)amino]benzaldehyde ≥98.0% (by total nitrogen basis)
Catalog # TCIAB3991-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:154.0 to 158.0 deg-C
Specification Test Results
About this item
:
:431.13 g/mol
CAS Number:25069-38-9
Formula:C₁₉H₁₃Br₂NO
MDL Number:MFCD12547118