Order Entry
Netherlands
ContactUsLinkComponent
4,4'-Bipiperidine ≥98.0% (by GC, titration analysis)
Irritant
4,4'-Bipiperidine ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Irritant
4,4'-Bipiperidine ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    169.0 to 173.0 deg-C

Specification Test Results

Frequently Bought Together