Order Entry
Netherlands
ContactUsLinkComponent
 
 
2,2'-Bipiperidine ≥98.0% (by titrimetric analysis)
  TCIAB3982-5G
 :  TCI
 :  
 
2,2'-Bipiperidine ≥98.0% (by titrimetric analysis)
  TCIAB3982-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 95.0 % (total of isomers)
  • Purity (Nonaqueous Titration):
    min. 98.0 %

 

 

 :
 :168.28 g/mol
 :93°C
 :208-514-9
 :GHS07
 :531-67-9
 :C₁₀H₂₀N₂
 :MFCD01572939