
Bleomycin sulphate (mixture) ≥85.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:9041-93-4
:
Formula:C₅₅H₈₄N₁₇O₂₁S₃·xH₂SO₄
MDL Number:MFCD00070310
:1512.6 g/mol
Merck Index:14,01318
EINECS:232-925-2
:GHS08
Storage Temperature:Koelkast
Specification Test Results
- Appearance:White to Orange to Green powder to crystaline
- Purity (HPLC):min.85.0 area% (Sum of A2,B2)
- BleomycinA2:55.0 to 70.0 area%
- BleomycinB2:25.0 to 32.0 area%
Specification Test Results
Frequently Bought Together