



Boron trichloride ∼13% ∼1.0 mol/L in p-xylene
TCIAB3968-100ML
: TCI
:
- Pk:100 mL
- Seq:2440
- Appearance:Light orange to Yellow to Green clear liquid
- Concentration (Neutralization titration):12.5 to 14.5 w/w%
:
:117.17 g/mol
:14,01348
:29°C
:Refrigerator
:233-658-4
:2920
:8,II
:GHS02
:GHS05
:GHS07
:GHS08
:10294-34-5
:BCl₃
:MFCD00011313