




Boron trichloride ∼16% ∼1.0 mol/L in heptane
Catalog # TCIAB3967-100ML
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Concentration (Neutralization titration):15.0 to 17.0 w/w%
Specification Test Results
About this item
:
:117.17 g/mol
Merck Index:14,01348
Flash Pt:2°C
Storage Temperature:Refrigerator
EINECS:233-658-4
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
:GHS09
CAS Number:10294-34-5
Formula:BCl₃
MDL Number:MFCD00011313