
2,2'-Bipyridine-6,6'-diol ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:103505-54-0
:MFCD18452865
Formula:C₁₀H₈N₂O₂
MDL Number:MFCD13194839
:188.19 g/mol
Melting Pt:340°C
:GHS07
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together