
5-Bromo-2-chloro-3-nitropyridine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:67443-38-3
:
Formula:C₅H₂BrClN₂O₂
MDL Number:MFCD00222270
:237.44 g/mol
:GHS07
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:66.0 to 70.0 deg-C
Specification Test Results
Frequently Bought Together