
(2-Bromophenyl)diphenylphosphine ≥95.0% (by GC)
Leverancier: TCI
Certificaten
Over dit artikel
CAS-nummer:62336-24-7
:
Formule:C₁₈H₁₄BrP
MDL-nummer:MFCD00045014
:341.19 g/mol
EINECS:263-515-1
:GHS07
Specificatie testresultaten
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:110.0 to 114.0 deg-C
Specificatie testresultaten
Frequently Bought Together