N-(tert-Butoxycarbonyl)-4-bromo-D-phenylalanine ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:79561-82-3
:
Formula:C₁₄H₁₈BrNO₄
MDL Number:MFCD00270359
:344.21 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Melting point:117.0 to 121.0 deg-C
- Specific rotation [a]20/D:-12.0 to -15.0 deg (C=1、MeOH)
Specification Test Results
Frequently Bought Together