4-tert-Butyl-N-(tert-butoxycarbonyl)-L-aspartate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1676-90-0
:
Formula:C₁₃H₂₃NO₆
MDL Number:MFCD00076912
:289.33 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:61.0 to 65.0 deg-C
- Specific rotation [a]20/D:+1.5 to +2.5 deg (c=1、MeOH)
Specification Test Results
Frequently Bought Together