Order Entry
Netherlands
ContactUsLinkComponent
3-Bromo-2-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
Irritant
3-Bromo-2-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Irritant
3-Bromo-2-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    168.0 to 172.0 deg-C

Specification Test Results

Frequently Bought Together