Order Entry
Netherlands
ContactUsLinkComponent
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid ≥97.0% (by HPLC)
Irritant
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid ≥97.0% (by HPLC)
Supplier:  TCI
Certificates
undefined
Irritant
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid ≥97.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Chelometric back titration):
    min. 90.0 %

Specification Test Results

Related Information

Frequently Bought Together