tert-Butyl-4-carbamoylpiperidine-1-carboxylate ≥97.0% (by GC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:91419-48-6
:
Formula:C₁₁H₂₀N₂O₃
MDL Number:MFCD02180953
:228.2900 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (with Total Nitrogen):min. 97.0 %
- Melting point:164.0 to 168.0 deg-C
Specification Test Results
Frequently Bought Together