
5-Bromo-2-chloronicotinic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAB3883-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:170.0 to 174.0 deg-C
Specification Test Results
About this item
:
:236.45 g/mol
:GHS07
CAS Number:29241-65-4
Formula:C₆H₃BrClNO₂
MDL Number:MFCD03844847