
1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid ≥95.0% (by GC, titration analysis)
Catalog # TCIAB3880-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Neutralization titration):min. 95.0 %
- Melting point:133.0 to 139.0 deg-C
Specification Test Results
About this item
:
:215.25 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:59378-75-5
Formula:C₁₀H₁₇NO₄
MDL Number:MFCD02179181