
2,3-Dihydrobenzo[b][1,4]dioxine-5-carboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4442-53-9
:
Formula:C₉H₈O₄
MDL Number:MFCD00239415
:180.16 g/mol
EINECS:224-670-0
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:190.0 to 194.0 deg-C
Specification Test Results
Frequently Bought Together