Order Entry
Netherlands
ContactUsLinkComponent
 
 
2,2'-Bipyridyl-1,1'-dioxide ≥98.0% (by HPLC, titration analysis)
  TCIAB3859-5G
 :  TCI
 :  
 
2,2'-Bipyridyl-1,1'-dioxide ≥98.0% (by HPLC, titration analysis)
  TCIAB3859-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %

 

 

 :
 :188.19 g/mol
 :311°C
 :GHS07
 :7275-43-6
 :C₁₀H₈N₂O₂
 :MFCD00065169