Order Entry
Netherlands
ContactUsLinkComponent
1,3-Bis[2-(4-aminophenyl)-2-propyl]benzene ≥98.0% (by GC, titration analysis)
Irriterend
1,3-Bis[2-(4-aminophenyl)-2-propyl]benzene ≥98.0% (by GC, titration analysis)
Catalog # TCIAB3853-25G
Leverancier:  TCI
CAS-nummer:  
Irriterend
1,3-Bis[2-(4-aminophenyl)-2-propyl]benzene ≥98.0% (by GC, titration analysis)
Catalog # TCIAB3853-25G
Leverancier:  TCI
CAS-nummer:  

Specificaties

  • Pk:
    25 g
  • Seq:
    3308

Specificaties

Specificatie testresultaten

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    113.0 to 117.0 deg-C

Specificatie testresultaten

Over dit artikel

 :
 :344.50 g/mol
 :GHS07
CAS-nummer:2687-27-6
Formule:C₂₄H₂₈N₂
MDL-nummer:MFCD00274255