


Bis(8-quinolinolato)copper(II) ≥98.0% (by titrimetric analysis), purified by sublimation
Catalog # TCIAB3834-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Dark green to Dark red to Black powder to crystal
- Purity (Chelometric Titration):min. 98.0 %
Specification Test Results
About this item
:
:351.85 g/mol
EINECS:233-841-9
UN:3077
ADR:9,III
:GHS07
:GHS08
:GHS09
CAS Number:10380-28-6
Formula:C₁₈H₁₂CuN₂O₂
MDL Number:MFCD00067392