BMB (1,4-Bis(maleimido)butane) ≥98.0% (by HPLC, total nitrogen)
: TCI
:28537-70-4
:
:C₁₂H₁₂N₂O₄
:MFCD01318342
:248.24 g/mol
:206°C
:GHS07
:Freezer
- Appearance:White to Light yellow to Dark green powder to crystaline
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Frequently Bought Together