



5-Bromo-5-nitro-1,3-dioxane ≥98.0% (by GC) for biochemical research
Catalog # TCIAB3769-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (GC):min. 98.0 %
- Melting point:59.0 to 63.0 deg-C
- Protein impurity test (SDS-PAGE):to pass test
Specification Test Results
About this item
:
:212.00 g/mol
Storage Temperature:Refrigerator
EINECS:250-001-7
UN:3261
ADR:8,I
:GHS05
:GHS07
:GHS08
:GHS09
CAS Number:30007-47-7
Formula:C₄H₆BrNO₄
MDL Number:MFCD00101855