

Benzyl-4-hydroxybenzoate ≥98.0% (by GC, titration analysis) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:94-18-8
:
Formula:C₁₄H₁₂O₃
MDL Number:MFCD00016471
:228.25 g/mol
EINECS:202-311-9
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:108.0 to 114.0 deg-C
- Protein impurity test (SDS-PAGE):to pass test
Specification Test Results
Frequently Bought Together