Order Entry
Netherlands
ContactUsLinkComponent
2,8-Bis(trifluoromethyl)quinolin-4-ol ≥98.0% (by GC)
Irritant
2,8-Bis(trifluoromethyl)quinolin-4-ol ≥98.0% (by GC)
Supplier:  TCI
Certificates
Irritant
2,8-Bis(trifluoromethyl)quinolin-4-ol ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    131.0 to 135.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together