3-Bromo-1H-pyrrolo[2,3-b]pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:74420-15-8
:
Formula:C₇H₅BrN₂
:197.04 g/mol
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:186.0 to 193.0 deg-C
Specification Test Results
Frequently Bought Together