N-(tert-Butoxycarbonyl)-4-nitro-D-phenylalanine ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:61280-75-9
:
Formula:C₁₄H₁₈N₂O₆
MDL Number:MFCD00069880
:310.31 g/mol
Melting Pt:115°C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 95.0 %
- Specific rotation [a]20/D:-7.0 to -10.0 deg (C=1, MeOH)
Specification Test Results
Frequently Bought Together