
trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:300364-84-5
:
Formula:C₁₇H₁₈N₂
MDL Number:MFCD03453260
:250.35 g/mol
UN:3439
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:123.0 to 127.0 deg-C
Specification Test Results
Frequently Bought Together