

2-(1,3-Benzodioxol-5-yl)-4,6-bis(trichloromethyl)-1,3,5-triazine ≥98.0% (by GC)
TCIAB3633-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:133.0 to 137.0 deg-C
:
:435.89 g/mol
:GHS07
:GHS08
:71255-78-2
:C₁₂H₅Cl₆N₃O₂
:MFCD04038074