N-(tert-Butoxycarbonyl)-3,4-difluoro-L-phenylalanine ≥95.0% (by HPLC, titration analysis)
Catalog # TCIAB3627-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Neutralization titration):min. 95.0 %
- Specific rotation [a]20/D:+4.0 to +6.5 deg (C=1,MeOH)
Specification Test Results
About this item
:
:301.29 g/mol
Melting Pt:83°C
CAS Number:198474-90-7
Formula:C₁₄H₁₇F₂NO₄
MDL Number:MFCD00672518