N-(tert-Butoxycarbonyl)-3,4-difluoro-D-phenylalanine ≥95.0% (by HPLC, titration analysis)
Catalog # TCIAB3626-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Neutralization titration):min. 95.0 %
Specification Test Results
About this item
:
:301.29 g/mol
CAS Number:205445-51-8
Formula:C₁₄H₁₇F₂NO₄
MDL Number:MFCD00672519