N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-L-alanine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:58438-04-3
:
Formula:C₁₈H₂₁NO₄
MDL Number:MFCD00079671
:315.37 g/mol
Melting Pt:100°C
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:+43.0 to +47.0 deg (C=1, EtOH)
Specification Test Results
Frequently Bought Together