N-(tert-Butoxycarbonyl)-3-(1-naphthyl)-D-alanine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB3615-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:+53.0 to +56.0 deg (C=2.5, MeOH)
Specification Test Results
About this item
:
:315.37 g/mol
CAS Number:76932-48-4
Formula:C₁₈H₂₁NO₄
MDL Number:MFCD00079668