

(S)-Benzyl-2-[4-(trifluoromethyl)phenyl]-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazolium tetrafluoroborate ≥97.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
:
Formula:C₁₉H₁₇BF₇N₃
ADR:8,III
:431.16 g/mol
UN:1759
:GHS05
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Optical purity (LC):min. 98.0 ee%
- Purity (with Total Nitrogen):min. 97.0 %
- Melting point:200.0 to 204.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together