
tert-Butyl-3-oxopyrrolidine-1-carboxylate ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:101385-93-7
:
Formula:C₉H₁₅NO₃
MDL Number:MFCD01631194
:185.22 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Gray to Brown powder to lump
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together