Order Entry
Netherlands
ContactUsLinkComponent
Benzoyl trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
Irritant
Benzoyl trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
Catalog # TCIAB3567-5G
Supplier:  TCI
CAS Number:  
Irritant
Benzoyl trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
Catalog # TCIAB3567-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :254.18 g/mol
Boiling Pt:94°C
Flash Pt:114°C
 :GHS07
Storage Temperature:Freezer
CAS Number:36967-85-8
Formula:C₈H₅F₃O₄S
MDL Number:MFCD17013536