
2-(2-Pyridyl)benzo[b]thiophene ≥97.0% (by HPLC, total nitrogen)
Catalog # TCIAB3561-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Melting point:123.0 to 127.0 deg-C
Specification Test Results
About this item
:
:211.28 g/mol
:GHS07
CAS Number:38210-35-4
Formula:C₁₃H₉NS
MDL Number:MFCD08457652