Order Entry
Netherlands
ContactUsLinkComponent
Bis(tetrabutylammonium)dihydrogenbis(isothiocyanato)bis(2,2'-bipyridyl-4,4'-dicarboxylato)ruthenium(II) ≥75.0% (by HPLC)
Health
Bis(tetrabutylammonium)dihydrogenbis(isothiocyanato)bis(2,2'-bipyridyl-4,4'-dicarboxylato)ruthenium(II) ≥75.0% (by HPLC)
Catalog # TCIAB3514-100MG
Supplier:  TCI
CAS Number:  
undefined
Health
Bis(tetrabutylammonium)dihydrogenbis(isothiocyanato)bis(2,2'-bipyridyl-4,4'-dicarboxylato)ruthenium(II) ≥75.0% (by HPLC)
Catalog # TCIAB3514-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Black powder to crystal
  • Purity (HPLC):
    min. 75.0 area%
  • Elemental analysis (Nitrogen):
    8.50 to 9.70 %

Specification Test Results

About this item

 :
 :1188.56 g/mol
 :GHS08
CAS Number:207347-46-4
Formula:C₅₈H₈₆N₈O₈RuS₂
MDL Number:MFCD11042475