Order Entry
Netherlands
ContactUsLinkComponent
2,2'-Bipyridine-5,5'-dicarboxylic acid ≥98.0% (by GC, titration analysis)
Irritant
2,2'-Bipyridine-5,5'-dicarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Irritant
2,2'-Bipyridine-5,5'-dicarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together