Order Entry
Netherlands
ContactUsLinkComponent
 
 
(1S,2R)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine ≥98.0% (by GC, titration analysis)
 :  TCI
 
 
(1S,2R)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 98.0 %
  • Optical purity (GC):
    min. 98.0 ee%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    -5.0 to -7.0 deg (C=1, CH2Cl2)

 

Frequently Bought Together