
Benzylhydrazine dihydrochloride ≥98.0% (by HPLC, titration analysis)
TCIAB3471-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Potassium iodate Method):min. 98.0 %
:
:195.09 g/mol
:145°C
:243-887-1
:2811
:6.1,III
:GHS06
:20570-96-1
:C₇H₁₀N₂
:MFCD00012921