
1-Bromo-3-phenylimidazo[1,5-a]pyridine ≥97.0% (by GC, titration analysis)
TCIAB3438-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Amber to Dark green powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
:
:273.13 g/mol
:GHS07
:104202-15-5
:C₁₃H₉BrN₂