Order Entry
Netherlands
ContactUsLinkComponent
Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV) 75% in isopropanol (IPA)
Flammable
Irritant
Health
Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV) 75% in isopropanol (IPA)
Supplier:  TCI
Certificates
Flammable
Irritant
Health
Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV) 75% in isopropanol (IPA)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Yellow to Orange clear liquid
  • Concentration (Gravimetric Method):
    75.0 to 80.0 w/w%

Specification Test Results

Frequently Bought Together