Order Entry
Netherlands
ContactUsLinkComponent
 
[1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene](1,3-diphenyl-1,3-propanedionato)copper(I) ≥98.0% (by titrimetric analysis)
 :  TCI
 
[1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene](1,3-diphenyl-1,3-propanedionato)copper(I) ≥98.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (Chelometric Titration):
    min. 98.0 %

 

Frequently Bought Together