Order Entry
Netherlands
ContactUsLinkComponent
N-(tert-Butoxycarbonyl)-4-fluoro-L-phenylalanine ≥98.0% (by titrimetric analysis)
N-(tert-Butoxycarbonyl)-4-fluoro-L-phenylalanine ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
N-(tert-Butoxycarbonyl)-4-fluoro-L-phenylalanine ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    83.0 to 87.0 deg-C
  • Specific rotation [a]20/D:
    +17.0 to +23.0 deg (C=1, EtOH)

Specification Test Results

Related Information

Frequently Bought Together