
tert-Butyl-2-(hydroxymethyl)pyrrolidine-1-carboxylate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:170491-63-1
:
Formula:C₁₀H₁₉NO₃
MDL Number:MFCD01456556
:201.3 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:52.0 to 56.0 deg-C
Specification Test Results
Frequently Bought Together