N-(tert-Butoxycarbonyl)-DL-2-phenylglycinol ≥93.0% (by GC)
: TCI
:67341-01-9
:
:C₁₃H₁₉NO₃
:MFCD01883039
:237.30 g/mol
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 93.0 %
- Melting point:141.0 to 147.0 deg-C
Frequently Bought Together