trans-4-(tert-Butoxycarbonylaminomethyl)cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:27687-14-5
:
Formula:C₁₃H₂₃NO₄
MDL Number:MFCD00673773
:257.33 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:133.0 to 137.0 deg-C
Specification Test Results
Frequently Bought Together