trans-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:53292-89-0
:
Formula:C₁₂H₂₁NO₄
MDL Number:MFCD01862293
:243.30 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:181.0 to 185.0 deg-C
Specification Test Results
Frequently Bought Together